Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001931.4 | NP_001922.2 | 9, 10 | 1957 | ||
GenBank mRNA | AK057299.1 | - | 9, 10 | 1957 | ||
AK223596.1 | - | 9, 10 | 1346 | |||
AK296274.1 | - | 9, 10 | 1236 | |||
AK297103.1 | - | 6, 7 | 703 | |||
AK299562.1 | - | 7, 8 | 794 | |||
AK303930.1 | - | 10, 11 | 1182 | |||
AL832703.1 | - | 9, 10 | 1328 | |||
BC039084.1 | - | 9, 10 | 1326 | 4508489 | ||
X13822.1 | - | 3, 4 | 290 | |||
Y00978.1 | - | 9, 10 | 1202 |