Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199196.1 | NP_001186125.1 | 4 | 655 | ||
NM_033415.3 | NP_219483.1 | 3 | 547 | |||
XM_005260157.3 | XP_005260214.1 | 3 | 598 | |||
XM_017027484.1 | XP_016882973.1 | 3 | 522 | |||
XM_017027485.1 | XP_016882974.1 | 5 | 779 | |||
GenBank mRNA | AK295488.1 | - | 3 | 321 | ||
BC056161.1 | - | 4 | 504 | |||
BC067125.1 | - | 4 | 424 | 6161112 | ||
BX648486.1 | - | 3 | 442 | |||
CX869901.1 | - | 4 | 655 | |||
Z36779.1 | - | 2 | 62 |