Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141968.1 | NP_001135440.1 | 14, 15 | 1194 | ||
NM_001271850.1 | NP_001258779.1 | 2, 3 | 473 | |||
NM_130785.3 | NP_570141.3 | 13, 14 | 1104 | |||
NM_199254.2 | NP_954863.2 | 16, 17 | 1335 | |||
NR_073485.1 | 18, 19 | 1447 | ||||
NR_073486.1 | 11, 12 | 774 | ||||
NR_073487.1 | 10, 11 | 872 | ||||
GenBank mRNA | AJ421032.1 | - | 13, 14 | 1104 | ||
AJ421033.1 | - | 10, 11 | 731 | |||
AK301741.1 | - | 18, 19 | 1447 | |||
AY219890.1 | - | 16, 17 | 1335 | |||
BC128146.1 | - | 14, 15 | 1176 | 40112547 | ||
BC128147.1 | - | 10, 11 | 854 | |||
FJ969729.1 | - | 2, 3 | 476 |
Gene Symbol: TPTE2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002793.1 | 19, 20 | 1978 |