Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPTE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141968.1 | NP_001135440.1 | 13, 14 | 1111 | ||
NM_001271850.1 | NP_001258779.1 | 1, 2 | 390 | |||
NM_130785.3 | NP_570141.3 | 12, 13 | 1021 | |||
NM_199254.2 | NP_954863.2 | 15, 16 | 1252 | |||
NR_073485.1 | 17, 18 | 1364 | ||||
NR_073486.1 | 10, 11 | 691 | ||||
NR_073487.1 | 9, 10 | 789 | ||||
GenBank mRNA | AJ421032.1 | - | 12, 13 | 1021 | ||
AJ421033.1 | - | 9, 10 | 648 | |||
AK301741.1 | - | 17, 18 | 1364 | |||
AY219890.1 | - | 15, 16 | 1252 | |||
BC128146.1 | - | 13, 14 | 1093 | 40112547 | ||
BC128147.1 | - | 9, 10 | 771 | |||
FJ969729.1 | - | 1, 2 | 393 |