Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR42E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145168.2 | NP_660151.2 | 3 | 521 | ||
XM_005256257.3 | XP_005256314.1 | 4 | 726 | |||
XM_011523471.2 | XP_011521773.1 | 3 | 535 | |||
XM_011523472.2 | XP_011521774.1 | 3 | 524 | |||
GenBank mRNA | AK026740.1 | - | 3 | 508 | ||
AK315652.1 | - | 3 | 482 | |||
BC019670.1 | - | 3 | 474 | 4517213 |