Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFAND4 SIRNA Assays ›
Gene Symbol:
ZFAND4
Gene Name:
zinc finger AN1-type containing 4
Gene Aliases:
ANUBL1
Chromosome Location:
Chr.10: 45614567 - 45672962 on Build GRCh38
UniGene ID:
Hs.659717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41158
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFAND4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128324.2
NP_001121796.1
7
1431
NM_001282905.1
NP_001269834.1
8
1881
NM_001282906.1
NP_001269835.1
8
1533
NM_174890.3
NP_777550.2
7
1779
XM_005271842.2
XP_005271899.1
6
1304
XM_005271843.2
XP_005271900.1
6
1471
XM_005271844.3
XP_005271901.1
4
937
XM_011540366.2
XP_011538668.1
5
1375
XM_011540368.1
XP_011538670.1
4
820
XM_017016931.1
XP_016872420.1
5
1395
XM_017016932.1
XP_016872421.1
7
1541
XM_017016933.1
XP_016872422.1
7
1408
XM_017016934.1
XP_016872423.1
7
1461
XM_017016935.1
XP_016872424.1
6
1211
XM_017016936.1
XP_016872425.1
5
1551
XM_017016937.1
XP_016872426.1
5
1399
XM_017016938.1
XP_016872427.1
5
1693
XM_017016939.1
XP_016872428.1
5
1264
XM_017016940.1
XP_016872429.1
8
1957
XM_017016941.1
XP_016872430.1
8
1500
XM_017016942.1
XP_016872431.1
6
1489
XM_017016943.1
XP_016872432.1
6
1765
XM_017016944.1
XP_016872433.1
6
1322
XM_017016945.1
XP_016872434.1
4
909
XM_017016946.1
XP_016872435.1
8
1575
GenBank mRNA
AF311324.1
-
7
1395
AK097504.1
-
4
899
AK292420.1
-
8
1482
AK292469.1
-
7
1380
AK314395.1
-
7
1391
BC045587.1
-
7
1431
5269483
BC048968.1
-
8
1423
5272133
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-4h6qx:80/100.66.76.46:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline