Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145088.1 | NP_001138560.1 | 14 | 1999 | ||
NM_145647.3 | NP_663622.2 | 14 | 1999 | |||
XM_005251102.1 | XP_005251159.1 | 14 | 1999 | |||
XM_005251103.1 | XP_005251160.1 | 14 | 1999 | |||
XM_011517378.2 | XP_011515680.1 | 14 | 2514 | |||
XM_011517379.2 | XP_011515681.1 | 14 | 1999 | |||
XM_017013983.1 | XP_016869472.1 | 14 | 1999 | |||
XM_017013984.1 | XP_016869473.1 | 13 | 2121 | |||
XM_017013985.1 | XP_016869474.1 | 13 | 1994 | |||
XM_017013986.1 | XP_016869475.1 | 13 | 1855 | |||
XM_017013988.1 | XP_016869477.1 | 13 | 1834 | |||
XM_017013989.1 | XP_016869478.1 | 13 | 1997 | |||
XM_017013990.1 | XP_016869479.1 | 12 | 1663 | |||
GenBank mRNA | AK056434.1 | - | 14 | 2000 | ||
AK094612.1 | - | 13 | 1797 | |||
BC027237.1 | - | 3 | 271 | |||
BC101724.1 | - | 14 | 1966 | 8069230 | ||
BC111954.1 | - | 14 | 1966 | 8327422 | ||
BC143523.1 | - | 13 | 1850 | |||
BC143525.1 | - | 14 | 1966 | 9052034 |