Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014765.1 | NP_001014765.1 | 4 | 788 | ||
NM_001304790.1 | NP_001291719.1 | 4 | 446 | |||
NM_001304791.1 | NP_001291720.1 | 3 | 527 | |||
NM_033182.5 | NP_149438.2 | 3 | 416 | |||
NM_183412.2 | NP_904319.1 | 3 | 527 | |||
NM_183413.2 | NP_904320.1 | 3 | 416 | |||
NR_130909.1 | 3 | 282 | ||||
XM_005263535.1 | XP_005263592.1 | 3 | 387 | |||
XM_005263536.4 | XP_005263593.1 | 4 | 781 | |||
XM_005263537.1 | XP_005263594.1 | 3 | 527 | |||
XM_006711043.2 | XP_006711106.1 | 3 | 387 | |||
XM_006711045.2 | XP_006711108.1 | 3 | 389 | |||
XM_011542435.1 | XP_011540737.1 | 4 | 849 | |||
XM_017002842.1 | XP_016858331.1 | 3 | 387 | |||
XM_017002843.1 | XP_016858332.1 | 3 | 389 | |||
XM_017002844.1 | XP_016858333.1 | 3 | 387 | |||
GenBank mRNA | AK055344.1 | - | 3 | 527 | ||
AK293721.1 | - | 3 | 364 | |||
AK294053.1 | - | 3 | 350 | |||
AK296025.1 | - | 3 | 262 | |||
AK301418.1 | - | 3 | 396 | |||
AK316284.1 | - | 3 | 396 | |||
AK316347.1 | - | 3 | 373 | |||
AY007380.1 | - | 2 | 323 | |||
AY040878.1 | - | 3 | 372 | |||
BC007832.2 | - | 3 | 363 | 4054414 | ||
BM546141.1 | - | 3 | 377 | |||
BM903606.1 | - | 423 | ||||
BU164151.1 | - | 4 | 370 | |||
CR749368.1 | - | 4 | 777 | |||
HY152948.1 | - | 3 | 415 |