Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272053.1 | NP_001258982.1 | 3 | 1884 | ||
NM_001272054.1 | NP_001258983.1 | 3 | 1931 | |||
NM_033296.2 | NP_150638.1 | 2 | 2003 | |||
GenBank mRNA | AA486335.1 | - | 1 | 157 | ||
AK057736.1 | - | 3 | 1357 | |||
AK074496.1 | - | 2 | 1410 | |||
AK123360.1 | - | 3 | 1884 | |||
AK129651.1 | - | 3 | 1288 | |||
AK303974.1 | - | 3 | 1330 | |||
AL539167.3 | - | 1 | 462 | |||
BG255189.1 | - | 1 | 166 | |||
BI858568.1 | - | 1 | 551 |