Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272053.1 | NP_001258982.1 | 3 | 1593 | ||
NM_001272054.1 | NP_001258983.1 | 3 | 1640 | |||
NM_033296.2 | NP_150638.1 | 2 | 1712 | |||
GenBank mRNA | AK057736.1 | - | 3 | 1066 | ||
AK074496.1 | - | 2 | 1119 | |||
AK123360.1 | - | 3 | 1593 | |||
AK129651.1 | - | 3 | 997 | |||
AK300242.1 | - | 3 | 1058 | |||
AK303974.1 | - | 3 | 1039 | |||
AL539167.3 | - | 1 | 171 | |||
BC022797.1 | - | 2 | 1098 | 5434657 | ||
BI858568.1 | - | 1 | 260 | |||
CB052459.1 | - | 1 | 479 |