Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007033.1 | NP_001007034.1 | 5 | 515 | ||
NM_001317999.1 | NP_001304928.1 | 4 | 425 | |||
GenBank mRNA | AK308011.1 | - | 5 | 583 | ||
AY321309.1 | - | 5 | 515 | |||
AY365135.1 | - | 5 | 460 | |||
BC132933.1 | - | 5 | 421 | 40146955 | ||
BC132935.1 | - | 5 | 421 | 40146957 |