Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 7 | 1242 | ||
NM_001172767.2 | NP_001166238.1 | 7 | 956 | |||
NM_014491.3 | NP_055306.1 | 7 | 1245 | |||
NM_148898.3 | NP_683696.2 | 8 | 1320 | |||
NM_148899.3 | NP_683697.2 | 6 | 881 | |||
NM_148900.3 | NP_683698.2 | 8 | 1296 | |||
NR_033766.1 | 11 | 1458 | ||||
NR_033767.1 | 8 | 1303 | ||||
XM_017012801.1 | XP_016868290.1 | 10 | 1926 | |||
GenBank mRNA | AF337817.1 | - | 7 | 1021 | ||
AF467253.1 | - | 6 | 883 | |||
AF467255.1 | - | 1 | 98 | |||
AF467257.1 | - | 7 | 923 | |||
AF467258.1 | - | 6 | 868 | |||
AF467259.1 | - | 7 | 943 | |||
AK131266.1 | - | 11 | 1442 | |||
AK296957.1 | - | 8 | 1176 | |||
AY144615.1 | - | 8 | 1315 | |||
BC126104.1 | - | 8 | 977 | 8991820 | ||
BC143866.1 | - | 8 | 977 | 9052382 | ||
BC143867.1 | - | 7 | 916 | 9052383 | ||
CR749236.1 | - | 13 | 1471 | |||
DQ778626.1 | - | 4 | 595 | |||
KJ534839.1 | - | 7 | 946 | |||
KJ535079.1 | - | 6 | 1084 | |||
KJ535086.1 | - | 6 | 870 |