Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 16 | 2326 | ||
NM_014491.3 | NP_055306.1 | 16 | 2329 | |||
NM_148898.3 | NP_683696.2 | 17 | 2404 | |||
NM_148900.3 | NP_683698.2 | 17 | 2380 | |||
NR_033766.1 | 19 | 2340 | ||||
NR_033767.1 | 17 | 2387 | ||||
XM_017012801.1 | XP_016868290.1 | 19 | 3010 | |||
GenBank mRNA | AF337817.1 | - | 16 | 2105 | ||
AK131266.1 | - | 19 | 2324 | |||
AK296957.1 | - | 17 | 2260 | |||
AY144615.1 | - | 17 | 2399 | |||
BC126104.1 | - | 17 | 2061 | 8991820 | ||
BC143866.1 | - | 17 | 2061 | 9052382 | ||
BC143867.1 | - | 16 | 2000 | 9052383 | ||
CR749236.1 | - | 23 | 2710 | |||
DQ778626.1 | - | 11 | 1400 | |||
KJ534839.1 | - | 16 | 2030 | |||
KJ535079.1 | - | 15 | 2168 | |||
KJ535086.1 | - | 15 | 1954 |