Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033198.3 | NP_149975.1 | 8 | 1256 | ||
GenBank mRNA | AB057723.1 | - | 8 | 883 | ||
AF161427.1 | - | 2 | 117 | |||
AK025825.1 | - | 1 | 21 | |||
AK075428.1 | - | 8 | 911 | |||
AK297046.1 | - | 7 | 775 | |||
AK314583.1 | - | 8 | 919 | |||
AY359112.1 | - | 7 | 1151 | |||
BC007301.1 | - | 6 | 640 | |||
BC069228.1 | - | 8 | 906 | 3944034 |