Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032796.3 | NP_116185.2 | 9 | 1139 | ||
NR_033181.1 | 9 | 1101 | ||||
GenBank mRNA | AF168955.1 | - | 9 | 1091 | ||
AF305828.1 | - | 6 | 631 | |||
AF338728.1 | - | 9 | 1091 | |||
AK027401.1 | - | 9 | 1081 | |||
AK126173.1 | - | 4 | 2054 | |||
AK222856.1 | - | 9 | 1073 | |||
AK295322.1 | - | 9 | 1076 | |||
AK303783.1 | - | 9 | 1055 | |||
AK314675.1 | - | 9 | 1111 | |||
BC001468.1 | - | 9 | 1070 | |||
BC014657.1 | - | 9 | 1059 | 4640905 | ||
CR749849.1 | - | 9 | 1125 |