Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032796.3 | NP_116185.2 | 3 | 491 | ||
NR_033181.1 | 3 | 453 | ||||
GenBank mRNA | AF168955.1 | - | 3 | 443 | ||
AF338728.1 | - | 3 | 443 | |||
AK027401.1 | - | 3 | 433 | |||
AK222856.1 | - | 3 | 425 | |||
AK295322.1 | - | 3 | 428 | |||
AK303783.1 | - | 3 | 407 | |||
AK314675.1 | - | 3 | 463 | |||
BC001468.1 | - | 3 | 422 | |||
BC014657.1 | - | 3 | 411 | 4640905 | ||
CR749849.1 | - | 3 | 476 | |||
DB047379.1 | - | 3 | 491 |