Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033194.2 | NP_149971.1 | 1 | 273 | ||
GenBank mRNA | AK093488.1 | - | 1 | 1581 | ||
AK130947.1 | - | 1 | 181 | |||
BC093991.1 | - | 1 | 194 | 7939836 | ||
BC093993.1 | - | 1 | 194 | 7939838 |