Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128150.1 | NP_001121622.1 | 4 | 455 | ||
NM_016467.4 | NP_057551.1 | 5 | 565 | |||
XM_005246970.2 | XP_005247027.1 | 4 | 484 | |||
XM_017005347.1 | XP_016860836.1 | 5 | 604 | |||
GenBank mRNA | AB064959.1 | - | 3 | 1024 | ||
AB064960.1 | - | 4 | 463 | |||
AF151036.1 | - | 4 | 403 | |||
AF395704.1 | - | 5 | 520 | |||
AK074756.1 | - | 4 | 454 | |||
AK075160.1 | - | 3 | 456 | |||
AK126336.1 | - | 1 | 3490 | |||
AK313533.1 | - | 5 | 530 | |||
BC005200.1 | - | 4 | 422 | 3929844 | ||
CR936635.1 | - | 3 | 1132 | |||
DB122266.1 | - | 4 | 455 |