Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320801.1 | NP_001307730.1 | 6 | 1861 | ||
NM_001320802.1 | NP_001307731.1 | 4 | 699 | |||
NM_001320803.1 | NP_001307732.1 | 4 | 413 | |||
NM_139280.3 | NP_644809.1 | 4 | 704 | |||
XM_017025345.1 | XP_016880834.1 | 4 | 557 | |||
XM_017025346.1 | XP_016880835.1 | 4 | 470 | |||
XM_017025347.1 | XP_016880836.1 | 4 | 650 | |||
XM_017025348.1 | XP_016880837.1 | 4 | 396 | |||
GenBank mRNA | AF373101.1 | - | 4 | 440 | ||
AF373102.1 | - | 4 | 510 | |||
AF395708.1 | - | 4 | 495 | |||
AK074811.1 | - | 4 | 501 | |||
AK075212.1 | - | 4 | 456 | |||
AK093063.1 | - | 6 | 1856 | |||
AX775843.1 | - | 4 | 458 | |||
AY357943.1 | - | 3 | 313 | |||
BC017087.1 | - | 4 | 470 | 3850211 | ||
BC071833.1 | - | 4 | 449 | 6340437 | ||
DB042662.1 | - | 4 | 414 | |||
DB127807.1 | - | 4 | 532 |