Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053045.1 | NP_444273.1 | 1 | 279 | ||
GenBank mRNA | AX775751.1 | - | 1 | 288 | ||
BC009283.2 | - | 1 | 279 | 4298098 | ||
BC009461.1 | - | 1 | 279 | 3538818 | ||
BC067786.1 | - | 1 | 291 | 5275460 |