Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163334.1 | NP_001156806.1 | 13 | 2007 | ||
NM_001163335.1 | NP_001156807.1 | 14 | 2119 | |||
NM_138780.2 | NP_620135.1 | 13 | 2028 | |||
XM_011544001.2 | XP_011542303.1 | 16 | 2446 | |||
XM_011544002.2 | XP_011542304.1 | 14 | 2080 | |||
XM_017029972.1 | XP_016885461.1 | 17 | 2612 | |||
XM_017029973.1 | XP_016885462.1 | 16 | 2679 | |||
XM_017029974.1 | XP_016885463.1 | 15 | 2307 | |||
GenBank mRNA | AB080222.1 | - | 12 | 1585 | ||
AK125106.1 | - | 13 | 1996 | |||
BC131585.1 | - | 13 | 1651 | 40113205 | ||
BX647688.1 | - | 14 | 2121 |