Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033226.2 | NP_150229.2 | 22 | 3523 | ||
GenBank mRNA | AF395908.1 | - | 22 | 3204 | ||
AF395909.1 | - | 21 | 2977 | |||
AF411577.1 | - | 22 | 3204 | |||
AF411578.1 | - | 22 | 3204 | |||
AY040220.1 | - | 22 | 3523 | |||
AY196326.1 | - | 20 | 3187 | |||
BC036378.1 | - | 23 | 3629 |