Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033226.2 | NP_150229.2 | 24, 25 | 3814 | ||
GenBank mRNA | AF395908.1 | - | 24, 25 | 3495 | ||
AF411577.1 | - | 24, 25 | 3495 | |||
AF411578.1 | - | 24, 25 | 3495 | |||
AK307109.1 | - | 2, 3 | 285 | |||
AK308599.1 | - | 2, 3 | 302 | |||
AY040220.1 | - | 24, 25 | 3814 | |||
AY196326.1 | - | 22, 23 | 3478 | |||
BC036378.1 | - | 25, 26 | 3920 | |||
DA096073.1 | - | 2, 3 | 302 |