Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033285.3 | NP_150601.1 | 3, 4 | 865 | ||
XM_011517386.2 | XP_011515688.1 | 3, 4 | 717 | |||
GenBank mRNA | AB017926.1 | - | 2, 3 | 520 | ||
AF409115.1 | - | 3, 4 | 837 | |||
AK315071.1 | - | 3, 4 | 807 | |||
BC074813.2 | - | 2, 3 | 513 | 30915317 | ||
BC074868.2 | - | 2, 3 | 513 | 30915467 |