Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257158.1 | NP_001244087.1 | 2 | 321 | ||
NM_001257159.1 | NP_001244088.1 | 2 | 321 | |||
NM_001257160.1 | NP_001244089.1 | 2 | 321 | |||
NM_018718.2 | NP_061188.1 | 2 | 321 | |||
NR_046443.1 | 2 | 321 | ||||
XM_011516709.2 | XP_011515011.1 | 2 | 168 | |||
GenBank mRNA | AF429308.1 | - | 2 | 167 | ||
AF429309.1 | - | 2 | 167 | |||
AJ278890.1 | - | 2 | 124 | |||
AK026098.1 | - | 2 | 121 | |||
AK298618.1 | - | 2 | 137 | |||
AK311382.1 | - | 2 | 138 | |||
AK314676.1 | - | 2 | 100 | |||
BC018789.1 | - | 2 | 103 | |||
BC056162.1 | - | 2 | 94 | 6142659 | ||
CB155730.1 | - | 2 | 348 | |||
DB069500.1 | - | 2 | 323 |