Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLSTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130749.1 | 1 | 386 | |||
GenBank mRNA | AW834946.1 | - | 1 | 457 | ||
S72422.1 | - | 1 | 384 |
Gene Symbol: DLST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244883.1 | NP_001231812.1 | 3, 4 | 212 | ||
NM_001933.4 | NP_001924.2 | 3, 4 | 212 | |||
NR_033814.1 | 3, 4 | 212 | ||||
GenBank mRNA | AK289414.1 | - | 3, 4 | 210 | ||
AK300396.1 | - | 3, 4 | 181 | |||
AK316463.1 | - | 3, 4 | 187 | |||
AU133381.1 | - | 3, 4 | 187 | |||
BC000302.2 | - | 3, 4 | 174 | 2820348 | ||
BC001922.1 | - | 3, 4 | 174 | 2820348 | ||
BX248276.1 | - | 3, 4 | 189 | |||
CR456727.1 | - | 3, 4 | 149 | |||
D16373.1 | - | 3, 4 | 161 | |||
DC338747.1 | - | 3, 4 | 213 | |||
L37418.1 | - | 3, 4 | 149 |