Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317902.1 | NP_001304831.1 | 2 | 644 | ||
NM_024831.7 | NP_079107.6 | 3 | 709 | |||
XM_005251328.2 | XP_005251385.1 | 2 | 424 | |||
XM_006716485.2 | XP_006716548.1 | 3 | 495 | |||
XM_017014004.1 | XP_016869493.1 | 3 | 495 | |||
GenBank mRNA | AF286340.1 | - | 3 | 799 | ||
AF432215.1 | - | 3 | 457 | |||
AK314693.1 | - | 3 | 483 | |||
AY028423.1 | - | 3 | 493 | |||
BX648548.1 | - | 2 | 407 | |||
DB082992.1 | - | 3 | 497 |