Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317902.1 | NP_001304831.1 | 7 | 2011 | ||
NM_024831.7 | NP_079107.6 | 8 | 2076 | |||
XM_005251328.2 | XP_005251385.1 | 7 | 1791 | |||
XM_006716485.2 | XP_006716548.1 | 8 | 1862 | |||
XM_017014004.1 | XP_016869493.1 | 8 | 1862 | |||
GenBank mRNA | AF286340.1 | - | 8 | 2166 | ||
AF432215.1 | - | 8 | 1824 | |||
AK055178.1 | - | 5 | 950 | |||
AK314693.1 | - | 8 | 1850 | |||
AL832426.1 | - | 2 | 332 | |||
AY534911.1 | - | 5 | 866 | |||
BX648548.1 | - | 7 | 1774 |