Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144944.1 | NP_001138416.1 | 3 | 355 | ||
NM_001144945.1 | NP_001138417.1 | 3 | 277 | |||
NM_033546.3 | NP_291024.1 | 3 | 336 | |||
GenBank mRNA | AK291726.1 | - | 3 | 316 | ||
AV714583.1 | - | 4 | 262 | |||
AY320408.1 | - | 3 | 340 | |||
BC004994.1 | - | 3 | 255 | 2906108 | ||
BG696903.1 | - | 3 | 356 | |||
BG703257.1 | - | 3 | 346 | |||
CN302451.1 | - | 3 | 258 | |||
D50372.1 | - | 3 | 289 | |||
D82058.1 | - | 3 | 269 | |||
U26162.1 | - | 3 | 310 |