Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144944.1 | NP_001138416.1 | 4 | 797 | ||
NM_001144945.1 | NP_001138417.1 | 4 | 719 | |||
NM_033546.3 | NP_291024.1 | 4 | 778 | |||
GenBank mRNA | AK291726.1 | - | 4 | 758 | ||
AY320408.1 | - | 4 | 782 | |||
BC004994.1 | - | 4 | 697 | 2906108 | ||
CN302451.1 | - | 4 | 700 | |||
D50372.1 | - | 4 | 730 | |||
D82058.1 | - | 4 | 711 | |||
U26162.1 | - | 4 | 752 |