Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320847.1 | NP_001307776.1 | 1, 2 | 315 | ||
NM_133491.4 | NP_597998.1 | 1, 2 | 315 | |||
GenBank mRNA | AF348524.1 | - | 1, 2 | 74 | ||
BC011751.2 | - | 1, 2 | 285 | 3534187 | ||
BG748008.1 | - | 1, 2 | 242 |