Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038493.1 | NP_001033582.1 | 2 | 542 | ||
NM_178120.4 | NP_835221.2 | 3 | 742 | |||
GenBank mRNA | AK095266.1 | - | 1 | 2520 | ||
AK290503.1 | - | 3 | 715 | |||
AY257976.1 | - | 3 | 742 | |||
BC036189.1 | - | 3 | 708 | 5311747 | ||
BC053351.1 | - | 2 | 419 | 5243874 | ||
KJ534824.1 | - | 3 | 544 |