Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052874.4 | NP_443106.1 | 4 | 511 | ||
XM_017022893.1 | XP_016878382.1 | 4 | 266 | |||
GenBank mRNA | AY028792.1 | - | 4 | 327 | ||
AY995211.1 | - | 4 | 269 | |||
BC038359.1 | - | 1 | 17 | |||
BC062298.1 | - | 4 | 323 | 6375788 | ||
DN990202.1 | - | 4 | 505 |