Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033448.2 | NP_258259.1 | 3 | 776 | ||
XM_017018749.1 | XP_016874238.1 | 4 | 649 | |||
GenBank mRNA | AJ308599.1 | - | 3 | 748 | ||
AJ308600.1 | - | 4 | 655 | |||
AJ308601.1 | - | 3 | 580 | |||
AK122795.1 | - | 3 | 776 | |||
BC103917.1 | - | 3 | 726 | 40006780 | ||
BC103918.1 | - | 3 | 726 | 40006781 |