Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282761.1 | NP_001269690.1 | 6 | 744 | ||
NM_080667.6 | NP_542398.3 | 5 | 669 | |||
XM_011532499.1 | XP_011530801.1 | 2 | 926 | |||
XM_017003254.1 | XP_016858743.1 | 4 | 545 | |||
XM_017003255.1 | XP_016858744.1 | 4 | 568 | |||
GenBank mRNA | AK309764.1 | - | 5 | 618 | ||
AY358097.1 | - | 6 | 736 | |||
BC001797.1 | - | 599 | ||||
BC010011.2 | - | 5 | 576 | 4309613 |