Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KTI12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138417.2 | NP_612426.1 | 1 | 781 | ||
GenBank mRNA | AF327348.1 | - | 1 | 762 | ||
AK313746.1 | - | 1 | 825 | |||
BC012173.1 | - | 1 | 745 | 4643409 |
Gene Symbol: TXNDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046405.1 | 3 | 2265 | |||
NR_046406.1 | 3 | 2142 |