Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324086.1 | NP_001311015.1 | 5 | 590 | ||
NM_001324410.1 | NP_001311339.1 | 5 | 491 | |||
NM_001324411.1 | NP_001311340.1 | 4 | 478 | |||
NM_001324412.1 | NP_001311341.1 | 4 | 392 | |||
NM_001324413.1 | NP_001311342.1 | 5 | 482 | |||
NM_001324414.1 | NP_001311343.1 | 5 | 538 | |||
NM_001324415.1 | NP_001311344.1 | 5 | 591 | |||
NM_058192.2 | NP_478072.1 | 5 | 581 | |||
GenBank mRNA | AL514840.3 | - | 5 | 441 | ||
AL520203.3 | - | 4 | 420 | |||
BC011783.2 | - | 4 | 446 | 3937184 | ||
BC012951.2 | - | 1 | 81 | |||
BC036279.2 | - | 1 | 81 | |||
BG686882.1 | - | 5 | 580 | |||
BX358716.2 | - | 4 | 377 | |||
CN311777.1 | - | 5 | 495 | |||
CN311779.1 | - | 5 | 440 | |||
CX786721.1 | - | 5 | 541 |