Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133373.4 | NP_588614.1 | 4 | 769 | ||
XM_011524253.2 | XP_011522555.1 | 4 | 754 | |||
GenBank mRNA | AB075844.1 | - | 4 | 551 | ||
AK074240.1 | - | 4 | 750 | |||
AL834392.1 | - | 4 | 611 | |||
BC010668.1 | - | 2 | 131 | |||
BC072384.1 | - | 4 | 734 | 6378601 |