Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_079834.2 | NP_524558.1 | 5 | 413 | ||
GenBank mRNA | AK074586.1 | - | 5 | 415 | ||
AK074967.1 | - | 5 | 388 | |||
AK307641.1 | - | 4 | 339 | |||
AK307700.1 | - | 4 | 337 | |||
AL832731.1 | - | 3 | 242 | |||
BC011747.2 | - | 5 | 375 | 3161979 | ||
BC016509.1 | - | 5 | 387 | 4180160 | ||
BC062598.1 | - | 5 | 386 | 5247983 |