Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AZIN2 SIRNA Assays ›
Gene Symbol:
AZIN2
Gene Name:
antizyme inhibitor 2
Gene Aliases:
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp
Chromosome Location:
Chr.1: 33081110 - 33162288 on Build GRCh38
UniGene ID:
Hs.101807
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41476
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AZIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001293562.1
NP_001280491.1
7, 8
1118
NM_001301823.1
NP_001288752.1
5, 6
1021
NM_001301824.1
NP_001288753.1
4, 5
847
NM_001301825.1
NP_001288754.1
5, 6
818
NM_001301826.1
NP_001288755.1
5, 6
818
NM_052998.3
NP_443724.1
8, 9
1333
NR_126031.1
5, 6
855
XM_005270404.2
XP_005270461.1
7, 8
1080
XM_005270406.1
XP_005270463.1
6, 7
947
XM_005270407.1
XP_005270464.1
5, 6
717
XM_006710312.1
XP_006710375.1
5, 6
765
XM_011540557.1
XP_011538859.1
6, 7
899
XM_011540563.1
XP_011538865.1
7, 8
1078
XM_011540566.1
XP_011538868.1
7, 8
1078
XM_017000167.1
XP_016855656.1
7, 8
1078
XM_017000168.1
XP_016855657.1
8, 9
1203
XM_017000169.1
XP_016855658.1
8, 9
1111
XM_017000170.1
XP_016855659.1
7, 8
1105
XM_017000171.1
XP_016855660.1
8, 9
1191
XM_017000172.1
XP_016855661.1
8, 9
1108
XM_017000173.1
XP_016855662.1
6, 7
784
XM_017000174.1
XP_016855663.1
7, 8
1078
XM_017000175.1
XP_016855664.1
6, 7
903
XM_017000176.1
XP_016855665.1
6, 7
877
XM_017000177.1
XP_016855666.1
6, 7
935
XM_017000178.1
XP_016855667.1
6, 7
888
GenBank mRNA
AB075825.1
-
4, 5
2936
AK057051.1
-
4, 5
821
AK095127.1
-
5, 6
1002
AK122697.1
-
4, 5
1726
AK315677.1
-
7, 8
1080
AL832640.2
-
7, 8
1869
AY050634.1
-
5, 6
746
AY050635.1
-
5, 6
746
AY050636.1
-
5, 6
782
AY050637.1
-
5, 6
746
AY325129.1
-
7, 8
1064
BC010449.1
-
7, 8
1064
4156927
BC028128.1
-
8, 9
1333
5164244
Back To Top
More Information
Panther Classification:
Molecular Function -
decarboxylase
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
putrescine transport
putrescine biosynthetic process from ornithine
negative regulation of protein catabolic process
positive regulation of catalytic activity
agmatine biosynthetic process
trans-Golgi network membrane organization
positive regulation of polyamine transmembrane transport
protein binding
arginine decarboxylase activity
putrescine transmembrane transporter activity
ornithine decarboxylase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline