Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173513.1 | NP_001166984.1 | 10 | 1410 | ||
NM_052883.1 | NP_443115.1 | 10 | 1410 | |||
GenBank mRNA | AF133519.1 | - | 10 | 1078 | ||
AF171055.1 | - | 10 | 1084 | |||
AK299502.1 | - | 10 | 1387 | |||
BC030028.2 | - | 10 | 1388 | |||
BC050032.1 | - | 10 | 1397 |