Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288585.1 | NP_001275514.1 | 2 | 468 | ||
NM_032857.4 | NP_116246.2 | 2 | 468 | |||
NM_171846.3 | NP_741982.1 | 2 | 468 | |||
XM_011521180.1 | XP_011519482.1 | 2 | 462 | |||
GenBank mRNA | AK027808.1 | - | 2 | 436 | ||
AY358709.1 | - | 2 | 416 | |||
BC067288.1 | - | 2 | 448 | 30366453 |