Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079821.2 | NP_001073289.1 | 4 | 441 | ||
NM_001127461.2 | NP_001120933.1 | 3 | 1083 | |||
NM_001127462.2 | NP_001120934.1 | 2 | 1049 | |||
NM_001243133.1 | NP_001230062.1 | 2 | 1049 | |||
NM_004895.4 | NP_004886.3 | 2 | 1049 | |||
NM_183395.2 | NP_899632.1 | 2 | 1049 | |||
XM_011544048.2 | XP_011542350.1 | 3 | 1188 | |||
XM_011544053.2 | XP_011542355.1 | 3 | 1076 | |||
XM_011544055.2 | XP_011542357.1 | 3 | 1076 | |||
XM_017000181.1 | XP_016855670.1 | 4 | 665 | |||
XM_017000182.1 | XP_016855671.1 | 4 | 610 | |||
XM_017000183.1 | XP_016855672.1 | 3 | 1076 | |||
XM_017000184.1 | XP_016855673.1 | 3 | 1076 | |||
GenBank mRNA | AB208891.1 | - | 2 | 1058 | ||
AF410477.1 | - | 2 | 1049 | |||
AF418985.2 | - | 2 | 465 | |||
AF420469.1 | - | 3 | 435 | |||
AF427617.1 | - | 2 | 1049 | |||
AF468522.1 | - | 2 | 465 | |||
AK027194.1 | - | 2 | 478 | |||
AK314998.1 | - | 4 | 441 | |||
AY092033.1 | - | 2 | 465 | |||
AY422168.1 | - | 2 | 297 | |||
BC117211.1 | - | 2 | 368 | 40125762 | ||
BC143359.1 | - | 2 | 368 | 9051864 | ||
BC143362.1 | - | 2 | 368 | 9051867 | ||
BC143363.1 | - | 2 | 368 | 9051868 | ||
DA244290.1 | - | 4 | 441 |