Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017922.1 | NP_001017922.1 | 4 | 642 | ||
NM_018538.3 | NP_061008.2 | 3 | 538 | |||
XM_006710313.3 | XP_006710376.1 | 5 | 907 | |||
XM_011540570.2 | XP_011538872.1 | 4 | 598 | |||
XM_011540571.2 | XP_011538873.1 | 4 | 626 | |||
GenBank mRNA | AF311285.1 | - | 3 | 538 | ||
AK056138.1 | - | 4 | 616 | |||
AK295704.1 | - | 4 | 560 | |||
AK314436.1 | - | 4 | 626 | |||
AY049028.1 | - | 3 | 556 | |||
BC099703.1 | - | 3 | 471 | 40000626 | ||
BC099707.1 | - | 3 | 476 | 40000628 | ||
BC099712.1 | - | 3 | 471 | 40000629 | ||
BC099713.1 | - | 3 | 471 | 40000630 | ||
BX537371.1 | - | 1 | 1140 |