Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134268.4 | NP_599030.1 | 4 | 1312 | ||
GenBank mRNA | AK098057.1 | - | 4 | 959 | ||
BC029798.1 | - | 4 | 988 | 5193583 |