Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMTK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080434.1 | NP_001073903.1 | 11, 12 | 1238 | ||
XM_011526411.2 | XP_011524713.1 | 11, 12 | 1569 | |||
XM_011526412.2 | XP_011524714.1 | 11, 12 | 1536 | |||
GenBank mRNA | AB067470.1 | - | 11, 12 | 1211 |