Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134707.1 | NP_001128179.1 | 7 | 1066 | ||
NM_007101.3 | NP_009032.2 | 7 | 1225 | |||
XM_017014367.1 | XP_016869856.1 | 7 | 1228 | |||
XM_017014368.1 | XP_016869857.1 | 7 | 1228 | |||
GenBank mRNA | AF047004.1 | - | 7 | 1064 | ||
AF047190.1 | - | 7 | 1064 | |||
AF095735.2 | - | 7 | 1190 | |||
AF095736.2 | - | 6 | 802 | |||
AF129265.1 | - | 3 | 218 | |||
AF162428.1 | - | 6 | 962 | |||
AJ223317.1 | - | 7 | 1064 | |||
AK223402.1 | - | 7 | 1112 | |||
AK291211.1 | - | 7 | 1060 | |||
AK298348.1 | - | 7 | 1109 | |||
AK316494.1 | - | 6 | 864 | |||
BC136363.1 | - | 7 | 1043 | 9020350 | ||
BC136364.1 | - | 7 | 1043 | 9020351 | ||
BC144035.1 | - | 7 | 1061 | 9052555 |