Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052900.2 | NP_443132.3 | 12 | 1784 | ||
NM_198123.1 | NP_937756.1 | 13 | 2096 | |||
NM_198124.1 | NP_937757.1 | 14 | 1988 | |||
XM_011516816.2 | XP_011515118.1 | 12 | 2270 | |||
XM_017013008.1 | XP_016868497.1 | 12 | 2504 | |||
XM_017013009.1 | XP_016868498.1 | 13 | 2582 | |||
XM_017013010.1 | XP_016868499.1 | 11 | 2192 | |||
XM_017013011.1 | XP_016868500.1 | 11 | 2192 | |||
XM_017013012.1 | XP_016868501.1 | 11 | 1555 | |||
GenBank mRNA | AB114604.1 | - | 13 | 2096 | ||
AB114605.1 | - | 14 | 1988 | |||
AY210419.1 | - | 12 | 1655 |