Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286250.1 | NP_001273179.1 | 6 | 1142 | ||
NM_001286251.1 | NP_001273180.1 | 5 | 1043 | |||
NM_001286252.1 | NP_001273181.1 | 7 | 1250 | |||
NM_001323252.1 | NP_001310181.1 | 8 | 1297 | |||
NM_001323253.1 | NP_001310182.1 | 7 | 1250 | |||
NM_001323254.1 | NP_001310183.1 | 6 | 1034 | |||
NM_001323255.1 | NP_001310184.1 | 6 | 1034 | |||
NM_001323256.1 | NP_001310185.1 | 8 | 1329 | |||
NM_001323257.1 | NP_001310186.1 | 6 | 1034 | |||
NM_001323258.1 | NP_001310187.1 | 8 | 2990 | |||
NM_001323259.1 | NP_001310188.1 | 7 | 1246 | |||
NM_001323260.1 | NP_001310189.1 | 7 | 1246 | |||
NM_001323261.1 | NP_001310190.1 | 5 | 760 | |||
NM_001323262.1 | NP_001310191.1 | 5 | 760 | |||
NM_001323263.1 | NP_001310192.1 | 5 | 764 | |||
NM_001323264.1 | NP_001310193.1 | 4 | 548 | |||
NM_052904.3 | NP_443136.2 | 7 | 1250 | |||
XM_005266813.4 | XP_005266870.1 | 5 | 655 | |||
XM_017010228.1 | XP_016865717.1 | 4 | 439 | |||
XM_017010229.1 | XP_016865718.1 | 5 | 612 | |||
GenBank mRNA | AB067487.1 | - | 5 | 664 | ||
AK055292.1 | - | 7 | 1136 | |||
AK295479.1 | - | 6 | 967 | |||
AK297240.1 | - | 5 | 787 | |||
BC071627.1 | - | 7 | 1125 | 5245540 | ||
DA255882.1 | - | 3 | 392 |