Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 18 | 2730 | ||
XM_005246263.2 | XP_005246320.1 | 18 | 2797 | |||
XM_005246265.3 | XP_005246322.1 | 18 | 2797 | |||
XM_011510530.1 | XP_011508832.1 | 19 | 2818 | |||
XM_011510531.1 | XP_011508833.1 | 19 | 2818 | |||
XM_011510532.2 | XP_011508834.1 | 19 | 2818 | |||
XM_011510533.2 | XP_011508835.1 | 19 | 2818 | |||
XM_011510534.1 | XP_011508836.1 | 19 | 2818 | |||
XM_011510535.2 | XP_011508837.1 | 18 | 2665 | |||
XM_011510536.2 | XP_011508838.1 | 13 | 1769 | |||
GenBank mRNA | AB067489.1 | - | 18 | 2501 | ||
AK094865.1 | - | 2 | 285 | |||
AK131316.1 | - | 4 | 928 | |||
AL834396.1 | - | 16 | 2114 | |||
BC113878.1 | - | 5 | 692 | |||
BC114438.1 | - | 5 | 697 | |||
BC167159.1 | - | 18 | 2501 | 8862581 |